Learn More
Kimberly-Clark Professional™ Guantes de nitrilo Kimtech Pure™ G3 y G5 STERLING™
Combinan la protección del nitrilo con la sensibilidad del látex
Marca:  Kimberly-Clark Professional™ 98186
Detalles adicionales : Peso : 9.90000kg
 Ver más versiones de este producto
Código de producto. 11792463
  • 281.52€ / paquete de 1500
Stock Remoto 11
Añadir a la cesta
Descripción
Descripción
Kimtech™ G5 Sterling™ Nitrile Gloves
  • Provide environmentally-friendly contamination control in a range of challenging cleanroom environments.
  • The gloves are made from 100% nitrile and are suitable for use in ISO 3 or higher cleanrooms.
  • The non-sterile gloves provide enhanced protection for both the wearer and their applications, and may be used in a range of processes and materials including semiconductors, non-sterile pharmaceuticals and medical devices.
  • Textured fingertips result in improved tactile control and sensitivity, while the powder-free static dissipative nitrile construction reduces electro-static effects for more stable precision use.
  • The gloves are ambidextrous and disposable
  • The efficient Sterling™ development process provides reduced thickness, meaning that not only is less force required to safely stretch the gloves, but storage space requirements and waste production are also greatly reduced.
  • The nitrile material reduces any risk of TYPE I glove-associated reactions relating to natural rubber latex.
  • Beaded cuffs give added strength and improved donning
  • Provided in double-bagged PE pouches making them cleanroom-ready.
  • The gloves are also designated as PPE Cat III according to (EU) Regulation 2016/425, and are certified for protection against viruses, micro-organisms and chemical splash.
  • Packaging: 6 bags x 250 ambidextrous, gray gloves (1,500 total)
Especificaciones
Especificaciones
ISO 3
Tacky
Ambidextrous
Nitrile
Powder-free
30 cm
Yes
Beaded
Yes
Medium
Gray
ISO 9001; CE CAT III; Category III PPE - CE Complex