missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15263-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The kinesins constitute a large family of microtubule-dependent motor proteins, which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. KIF1B is a member of the KIF1/Unc104 family of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1B is an amino-terminal-type motor protein that is ubiquitously expressed, with the most abundant levels in differentiated nerve cells. The human KIF1B gene maps to chromosome 1p36.2. Defects in axonal transport due to mutations at the KIF1B gene can underlie the human peripheral neuropathy phenotype. The mouse KIF1B gene generates an alternatively spliced transcript, which produces two isoforms.Especificaciones
KIF1B | |
Polyclonal | |
Unconjugated | |
KIF1B | |
CMT2, CMT2A, CMT2A1, HMSNII, KIAA0591, KIAA1448, KIF1B, kinesin family member 1B, Kinesin like protein KIF1B, KLP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117548, 16561, 23095 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60333, O88658, Q60575 | |
Kif1b | |
KIF1B Fusion Protein Ag7263 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |