missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1429 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25712-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.Especificaciones
KIAA1429 | |
Polyclonal | |
Unconjugated | |
KIAA1429 | |
fSAP121, KIAA1429, MSTP054, Protein virilizer homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25962 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q69YN4 | |
KIAA1429 | |
KIAA1429 Fusion Protein Ag22449 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |