missing translation for 'onlineSavingsMsg'
Learn More

KHK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15681-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814964

  • 109.00€ / 20 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones

Especificaciones

KHK
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50053, P97328, Q02974
Hepatic fructokinase, Ketohexokinase, ketohexokinase (fructokinase), KHK
Rabbit
IgG
20 μL
-20°C
Primary
16548, 25659, 3795
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
KHK
Liquid
Khk
KHK Fusion Protein Ag8182
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados