Learn More
Keratinocyte-SFM Medium (Kit) with L-glutamine, EGF, and BPE
Product Code. 10144892 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
10144892 1 Kit 1 set
1 options
This item is not returnable. View return policy
Product Code. 10144892 Supplier Gibco Supplier No. 17005075
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
with L-Glutamine, EGF and BPE.
Shelf life after supplementation: 2 weeks
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Keratinocyte Cells, Epithelial Cells
Classification Serum-free
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red, EGF, BPE
Quantity 1 Kit
pH 7 to 8.1
Content And Storage • 500 ml 1 X Keratinocyte-SFM with L-glutamine (store at 2°C to 8°C)
• 1 vial Human Recombinant EGF (store at -5°C to -30°C)
• 1 vial Bovine Pituitary Extract (store at -5°C to -30°C)
Culture Type Mammalian Cell Culture
Show More Show Less
For how long can I store Keratinocyte-SFM at 4 degrees C?
The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C
For Research Use Only. Not for use in diagnostic procedures.