Learn More
KDEL Antibody (1F2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-61924B
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213534
  • 623.09€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
KDEL Monoclonal specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones
Especificaciones
KDEL
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
KDELC1
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA)
100 μL
Primary
Detects KDEL tagged proteins.
Store at 4C in the dark.
IgG1
Western Blot
1F2
Western Blot
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302
Mouse
Protein G purified
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
79070
Human, Mouse, Rat
Purified