Learn More
KDEL Polyclonal Antibody, Alexa Fluor™ 647, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA1013A647
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1013A647 detects KDEL from human, rat, mouse and hamster. The PA1013A647 immunogen is a synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94.
KDEL Polyclonal antibody specifically detects KDEL in Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry.Especificaciones
KDEL | |
Polyclonal | |
Alexa Fluor 647 | |
Hsp90b1 | |
78 kDa glucose-regulated protein; 8030486F04Rik; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; AL022860; AU019543; AW215843; baffled; Binding-immunoglobulin protein; BiP; D2Wsu141e; D2Wsu17e; ECGP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; endoplasmic reticulum resident protein 58; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endothelial cell (HBMEC) glycoprotein; EP58; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; ER protein 58; ERD2; Erd2.1; ERp58; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein, 78kDa; gp96; gp96 homolog; GRP 78; Grp78; GRP-78; GRP94; GRP-94; HDEL; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kD protein 5 (gl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100689305, 100750912, 100773331, 10945, 14828, 22027, 25617, 3309, 361577, 362862, 68137, 7184 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P06761, P08113, P11021, P14625, P20029, P24390, Q569A6, Q66HD0, Q99JH8 | |
Hsp90b1, HSPA5, Kdelr1 | |
Synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94 | |
50 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |