missing translation for 'onlineSavingsMsg'
Learn More

KCTD15 Antibody (AT4C3), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-49429JF549

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213572

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

KCTD15
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
KCTD15
Recombinant human KCTD15 (1-234aa) purified from E. coli.
0.1 mL
79047
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
4C3
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
Mouse
Protein G purified
Primary
Human KCTD15 (1-234aa)
Store at 4°C in the dark.
IgG3 λ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales