Learn More
Novus Biologicals™ KCTD10 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-12207
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213333
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
The KCTD10 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC27723 and based on accession number NM_31954. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens potassium channel tetramerisation domain containing 10 (KCTD10), mRNA.
Native