missing translation for 'onlineSavingsMsg'
Learn More

KCNK10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87663

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215437

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

KCNK10 Polyclonal specifically detects KCNK10 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

KCNK10
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Protein A purified
RUO
54207
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
1 mg/ml
Western Blot 1.0 ug/ml
2P domain potassium channel TREK2, K2p10.1, Outward rectifying potassium channel protein TREK-2, potassium channel subfamily K member 10, potassium channel TREK-2, potassium channel, subfamily K, member 10, TREK-2, TREK-2 K(+) channel subunit, TREK2FLJ43399, TWIK-related K+ channel 2
The immunogen is a synthetic peptide directed towards the C-terminal region of human KCNK10
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales