Learn More
Kir3.4 (KCNJ5) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553594
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: VTPWDPKKIP KQARDYVPIA TDRTRLLAEG KKPRQRYMEK SGKCNVHHGN VQETY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Kir3.4 (KCNJ5) Polyclonal antibody specifically detects Kir3.4 (KCNJ5) in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
Kir3.4 (KCNJ5) | |
Polyclonal | |
Unconjugated | |
KCNJ5 | |
cardiac ATP-sensitive potassium channel; cardiac inward rectifier; CIR; G protein-activated inward rectifier potassium channel 4; GIRK4; GIRK-4; Heart KATP channel; inward rectifier K(+) channel Kir3.4; inward rectifier K+ channel KIR3.4; IRK4; IRK-4; KATP1; KATP-1; KCNJ5; Kir3.4; LQT13; Potassium channel, inwardly rectifying subfamily J member 5; potassium channel, inwardly rectifying subfamily J, member 5; potassium inwardly-rectifying channel, subfamily J, member 5; potassium voltage-gated channel subfamily J member 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48544 | |
KCNJ5 | |
Recombinant protein corresponding to Human KCNJ5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only