missing translation for 'onlineSavingsMsg'
Learn More

KCNIP4 Mouse anti-Human, Porcine, Rat, Clone: 5B1D12, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60133-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16827105

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.
Especificaciones

Especificaciones

KCNIP4
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6PIL6, Q99MG9
KCNIP4
KCNIP4 Fusion Protein Ag4863
20 μL
Primary
Human, Porcine, Rat
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
5B1D12
Unconjugated
KCNIP4
CALP, Calsenilin like protein, KCHIP4, KCNIP4
Mouse
Caprylic Acid Precipitation
RUO
100516499, 259243, 80333
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales