missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kanamycin, 50 mg/ml in distilled water, sterile-filtered

Código de producto. 17684465
Cambiar vista
Click to view available options
Cantidad:
50 mL
Tamaño de la unidad:
50 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
17684465 50 mL 50 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17684465 Proveedor Thermo Scientific Chemicals N.º de proveedor J67354.AD
  • ONLINE EXCLUSIVE PRICE
    247.86€
    205.65€ / 50 ml
    Ahorro 42.21€
    17% Descuento
 Artículo restringido
En stock
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

It has been used in tissue culture media. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. It can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL. Kanamycin can be used as a selection agent in several types of isolation media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Kanamycin monosulfate is the sulfate salt form of kanamycin A
• It was isolated from Streptomyces kanamyceticus
• This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

• Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
• This compound is also involved in translating prokaryotic ribosomes
• In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 25389-94-0
Fórmula molecular C18H38N4O15S
Peso molecular (g/mol) 582.575
Número MDL MFCD00070289
Clave InChI OOYGSFOGFJDDHP-IZQIRFRQSA-N
PubChem CID 134129479
Nombre IUPAC (2R,3S,4S,5S,6R)-2-(aminometilo)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihidroxi-6-(hidroximetil)oxan-2-il]oxi-2-hidroxiciclohexil]oxioxano-3,4,5-triol; ácido sulfúrico
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Especificaciones

Nombre del producto químico o material Kanamycin
Tipo Kanamycin
Nombre de nota 50mg/mL in distilled water, sterile-filtered
Forma física Liquid
Cantidad 50 mL
Información de solubilidad Fully miscible in water. Insoluble in acetone,chloroform,ether,ethyl acetate,and alcohol.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.