missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KALRN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19740-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody can recognize isoform1 and isoform2 of KALRN.
HAP1 (huntingtin-associated protein 1) binds to huntingtin. Huntingtin is a protein that contains a polyglutamine region and when the number of glutamine repeats exceeds 35, the gene encodes a version of huntingtin that leads to Huntington′s disease (HD). The ability of HAP1 to bind to huntingtin is enhanced by an expanded polyglutamine repeat region. HAP1 shows neuronal localization and moves with huntingtin in nerve fibers. HAP1 is primarily expressed in brain tissue, with greater expression in the olfactory bulb and brain stem. Mouse HAP1 is localized to membrane-bound organelles including large endosomes, tubulovesicular structures and budding vesicles in neurons. Duo, also designated huntingtin-associated protein interacting protein or HAPIP, binds Huntingtin-associated protein 1 (HAP1) and may have a role in vesicle trafficking and cytoskeletal function.Especificaciones
KALRN | |
Polyclonal | |
Unconjugated | |
KALRN | |
DUET, duo, HAPIP, Kalirin, kalirin, RhoGEF kinase, KALRN, Protein Duo, TRAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
545156, 8997 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2CG49, O60229 | |
Kalrn | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |