missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 8/Neuropsin Antibody (140), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-89988AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213836

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Kallikrein 8/Neuropsin Monoclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

Kallikrein 8/Neuropsin
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot
140
Western Blot
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 8/Neuropsin (O60259-1; Met1-Gly260).
0.1 mL
Neuroscience
11202
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales