missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10815-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817453

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
Especificaciones

Especificaciones

Kallikrein 1
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P06870
hK1, kallikrein 1, KLK1, KLKR, Tissue kallikrein
Rabbit
IgG
20 μL
-20°C
Primary
3816
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
KLK1
Liquid
KLK1
Kallikrein 1 Fusion Protein Ag1119
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados