Learn More
Invitrogen™ Human JMJD2A Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0920
Detalles adicionales : Peso : 0.01000kg
Descripción
JMJD2A Synthetic Peptide, PEP-0920, from Invitrogen.
This peptide corresponds to 14 amino acids near the center of human JMJD2A.PEP-0920 can be used as a blocking peptide with polyclonal antibody PA5-20806.Especificaciones
9682 | |
purified | |
Human JMJD2A | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O75164 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
KDM4A | |
0.02% sodium azide | |
50 μg | |
PBS |