Learn More
Invitrogen™ JAK3 Recombinant Human Protein
Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family.
Marca: Invitrogen™ PR7507B
Detalles adicionales : Peso : 0.01000kg
Descripción
JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors.
Especificaciones
NP_000206 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
JAK3, LJAK, JAKL | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
67.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
JAK Kinase Family | |
Recombinant | |
Baculovirus |