missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ JAK3 (Human) Recombinant Protein
Human JAK3 full-length ORF ( AAH28068, 1 a.a. - 619 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003718-P01.2ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAPPSEETPLIPQRSCSLLSTEAGALHVLLPARGPGPPQRLSFSFGDHLAEDLCVQAAKASGILPVYHSLFALATEDLSCWFPPSHIFSVEDASTQVLLYRIRFYFPNWFGLEKCHRFGLRKDLASAILDLPVLEHLFAQHRSDLVSGRLPVGLSLKEQGECLSLAVLDLARMAREQAQRPGELLKTVSYKACLPPSLRDLIQGLSFVTRRRIRRTVRRALRRVAACQADRHSLMAKYIMDLERLDPAGAAETFHVGLPGALGGHDGLGLLRVAGDGGIAWTQGEQEVLQPFCDFPEIVDISIKQAPRVGPAGEHRLVTVTRTDNQILEAEFPGLPEALSFVALVDGYFRLTTDSQHFFCKEVAPPRLLEEVAEQCHGPITLDFAINKLKTGGSRPGSYVLRRSPQDFDSFLLTVCVQNPLGPDYKGCLIRRSPTGTFLLVGLSRPHSSLRELLATCWDGGLHVDGVAVTLTSCCIPRPKEKSNLIVVQRGHSPPTSSLVQPQSQYQLSQMTFHKIPADSLEWHENLGHGSFTKIYRGCRHEVVDGEARKTEVLLKVMDAKHKNCMESFLEAASLMSQVSYRHLVLLHGVCMAGDSESPPPTHPTPASPKSRLFFPPLF
Especificaciones
AAH28068 | |
Janus kinase 3 (a protein tyrosine kinase, leukocyte) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JAK3 | |
GST |
3718 | |
Wheat germ expression system | |
2 μg | |
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |