Learn More
Hughes Jacketed Mobile Safety Shower with Eye/Face Wash
Product Code. 17752348
Change view
Click to view available options
Color:
White
Unit Size:
Each
Product Code. Color Unit Size
17752348 White Each
1 options
This item is not returnable. View return policy
Product Code. 17752348 Supplier Hughes Supplier No. HSHJ040KGW
  • ONLINE EXCLUSIVE PRICE
    8481.93€
    5505.66€ / paquete de 1
    Ahorro 2976.27€
    35% Descuento
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
114 L insulated mobile self-contained emergency safety shower with eye/face wash unit. Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tires, making it easy to maneuver by one person.
  • 114 liter capacity insulated mobile self-contained emergency safety shower with eye/face wash unit
  • Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tyres, making it easy to manoeuvre by one person
  • For long distances or uneven surfaces an optional 'A' frame with towing hitch can be fitted
  • In addition, the wheel spacing can be altered allowing the unit to fit through standard door openings
Specifications
Capacity (Metric) 114 L
Color White
Product Type Safety Shower