Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
Product Code. 15884744
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15884744 500 mL Each
15854744 25 mL Each
15874744 50 mL Each
15834744 100 mL Each
15864744 250 mL Each
15844744 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15884744 Supplier GPE Scientific™ Supplier No. CSY/500
Fecha estimada de envoi: 09-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Includes O-ring tap
  • Includes cold finger plain arm
Specifications
Capacity (Metric) 500 mL
Includes O-ring tap and cold finger plain arm
Product Type Sample Flask