Learn More
STEMCELL Technologies IWP-4
WNT pathway inhibitor; Inhibits Porcupine
Marca:  STEMCELL Technologies 72552
Detalles adicionales : Peso : 0.05000kg
 Ver más versiones de este producto
Código de producto. 17180082
  • 77.14€ / 1 miligramo
Fecha estimada de envío: 04-04-2024
para ver el stock
Descripción
Descripción
  • Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.) IWP-4 impairs WNT pathway activity in vitro with an IC50 value of 25 nM (Chen et al.)
  • Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al.), 2012, 2013; Sequiera et al.)
  • Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.)
Especificaciones
Especificaciones
686772-17-8
1 mg
≥95%
Inhibitor of Wnt Production-4
C23H20N4O3S3