missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ITK, GST Tag Recombinant Protein
ITK Recombinant Human Protein
Marca: Invitrogen PV3875
Detalles adicionales : Peso : 0.05250kg
Descripción
ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation.Especificaciones
NP_005537 | |
114294, 3702 | |
10μg | |
TEC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
IL2-inducible T-cell kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
99.9 kDa | |
ITK, EMT, LYK, PSCTK2 | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
ITK | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |
For Research Use Only. Not for use in diagnostic procedures.