missing translation for 'onlineSavingsMsg'
Learn More

ITIH3 Antibody (049), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90329V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213639

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

ITIH3 Monoclonal specifically detects ITIH3 in Human samples. It is validated for ELISA.
Especificaciones

Especificaciones

ITIH3
Monoclonal
DyLight 405
50mM Sodium Borate
Rabbit
Protein A purified
RUO
3699
Store at 4°C in the dark.
IgG
ELISA
49
ELISA
H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP
This antibody was obtained from a rabbit immunized with purified, recombinant Human ITIH3 (NP_002208.3; Met1-Asp651).
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales