Learn More
ISPD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560520
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: SELNHVKVTS EALGHAGRHL QQIILDQCYN FVCVNVTTSD FQETQKLLSM LEESSLCILY PVVVVSVHFL DFKLVPPSQK MENLMQ Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 58%.
ISPD Polyclonal antibody specifically detects ISPD in Human samples. It is validated for Immunohistochemistry.Especificaciones
ISPD | |
Polyclonal | |
Unconjugated | |
ISPD | |
2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4930579E17Rik; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; AV040780; Crppa; D-ribitol-5-phosphate cytidylyltransferase; hCG_1745121; hISPD; isoprenoid synthase domain containing; isoprenoid synthase domain-containing protein; Ispd; MDDGA7; MDDGC7; Nip; notch1-induced protein; testicular tissue protein Li 97 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
729920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A4D126 | |
ISPD | |
Recombinant protein corresponding to Human ISPD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only