Learn More
ISOC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567096
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: HIVADATSSRS MMDRMFALER LARTGIIVTT SEAVLLQLVA DKDHPKFKEI QNLIKASAPE SGLLSKV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
ISOC1 Polyclonal antibody specifically detects ISOC1 in Human samples. It is validated for Immunocytochemistry.Especificaciones
ISOC1 | |
Polyclonal | |
Unconjugated | |
ISOC1 | |
2610034N03Rik; CGI-111; Down-regulated in nephrectomized rat kidney #1; down-regulated in nephrectomized rat kidney protein 1; DR-NR#1; DR-NR1; isoc1; isochorismatase domain containing 1; isochorismatase domain-containing protein 1; isochorismatase domain-containing protein 1; LOW QUALITY PROTEIN: isochorismatase domain-containing protein 1; RGD1307632; zgc:153699 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CN7 | |
ISOC1 | |
Recombinant Human ISOC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only