Learn More
Isobutiramida, 99 %, Thermo Scientific Chemicals
Descripción
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble en agua.
Notas
Incompatible con ácidos, bases, agentes oxidantes fuertes y agentes reductores fuertes.
Especificaciones
Nombre del producto químico o material | Isobutyramide |
Punto de fusión | 127°C to 129°C |
Densidad | 1.013 |
Punto de ebullición | 216°C to 220°C |
Cantidad | 100 g |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 87.12 |
Porcentaje de pureza | 99% |
RUO – Research Use Only