missing translation for 'onlineSavingsMsg'
Learn More

Isobutiramida, 99 %, Thermo Scientific Chemicals

Catalog Number p-7025911
Click to view available options
Cantidad:
25 g
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
563-83-7
C4H9NO
87.122
MFCD00008019
WFKAJVHLWXSISD-UHFFFAOYSA-N
isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
68424
2-metilpropanamida
CC(C)C(=O)N
Este artículo no se puede devolver. Vea la política de devoluciones
563-83-7
C4H9NO
87.122
MFCD00008019
WFKAJVHLWXSISD-UHFFFAOYSA-N
isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
68424
2-metilpropanamida
CC(C)C(=O)N

Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubilidad
Soluble en agua.

Notas
Incompatible con ácidos, bases, agentes oxidantes fuertes y agentes reductores fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Isobutyramide
Punto de fusión 127°C to 129°C
Densidad 1.013
Punto de ebullición 216°C to 220°C
Cantidad 25 g
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 87.12
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'