Learn More
Brady™ ISO 7010 Sign - Wear eye protection
Product Code. 15690785 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15690785 200 mm Each
15634457 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15690785 Supplier Brady™ Supplier No. 818425
  • ONLINE EXCLUSIVE PRICE
    13.60€
    13.34€ / paquete de 1
    Ahorro 0.26€
    1% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M004-Dia 200-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
Diameter (Metric) 200 mm
For Use With (Application) Indoor and Outdoor
Thickness 1.501 mm
Temperature Range (Metric) -40°C to 120°C
Shape Round
Languages Global
Color White on Blue