missing translation for 'onlineSavingsMsg'
Learn More

ISG15/UCRP Antibody (239), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90146F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213356

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

ISG15/UCRP
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA, Immunofluorescence
239
ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence
G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17
This antibody was obtained from a rabbit immunized with purified, recombinant Human ISG15/UCRP (AAH09507.1; Met1-Ser165).
0.1 mL
Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway
9636
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales