missing translation for 'onlineSavingsMsg'
Learn More

sal de amonio del ácido isetiónico, 99 %, Thermo Scientific™

Catalog Number p-7046447
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
57267-78-4
C2H9NO4S
143.157
MFCD00012847
LLOHIFXFHGMBNO-UHFFFAOYSA-N
ammonium isethionate, ammonium 2-hydroxyethanesulphonate, ammonium 2-hydroxyethanesulfonate, isethionic acid ammonium salt, ethanesulfonic acid, 2-hydroxy-, monoammonium salt, 2-hydroxyethanesulfonic acid ammonium salt, 2-hydroxyethanesulfonic acid, monoammonium salt, ethanesulfonic acid, 2-hydroxy-, ammonium salt 1:1, ethanesulfonic acid, 2-hydroxy-, ammonium salt, acmc-1axee
93602
azanio;2-hidroxietanosulfonato
C(CS(=O)(=O)[O-])O.[NH4+]
Este artículo no se puede devolver. Vea la política de devoluciones
57267-78-4
C2H9NO4S
143.157
MFCD00012847
LLOHIFXFHGMBNO-UHFFFAOYSA-N
ammonium isethionate, ammonium 2-hydroxyethanesulphonate, ammonium 2-hydroxyethanesulfonate, isethionic acid ammonium salt, ethanesulfonic acid, 2-hydroxy-, monoammonium salt, 2-hydroxyethanesulfonic acid ammonium salt, 2-hydroxyethanesulfonic acid, monoammonium salt, ethanesulfonic acid, 2-hydroxy-, ammonium salt 1:1, ethanesulfonic acid, 2-hydroxy-, ammonium salt, acmc-1axee
93602
azanio;2-hidroxietanosulfonato
C(CS(=O)(=O)[O-])O.[NH4+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Isethionic acid ammonium salt
Punto de fusión 138°C to 139°C
Cantidad 1 g
Beilstein 3722656
Sensibilidad Hygroscopic
Formula Weight (peso de la fórmula) 143.16
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'