Learn More
Perclorato de hierro(III) hidrato, agregados cristalinos blancos, Thermo Scientific Chemicals
Descripción
Iron(III) perchlorate hydrate is used as a starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units that allow two coordination modes. It is also used as a catalyst for synthesis of fullerene-fused lactones, α-Carbonyl furans via one-pot cyclization, fullerodioxolanes via heterocyclization, dialylated indoles via double alkylation, aryl esters via oxidative esterification and as a chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution, as an oxidant for conducting polymer nanoparticles synthesized in ionic liquid by chemical polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Iron(III) perchlorate hydrate |
Color | Blanco |
Forma física | cristalino/a/s |
Cantidad | 500 g |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 354.19 (Anhydrous) |
Grado | Reactivo |
Olor | Odorless |
Sensibilidad | Hygroscopic |
RUO – Research Use Only