Learn More
Hierro, 99,99 %, (base de trazas metálicas), partículas, Thermo Scientific Chemicals
Catalog Number p-4228260
Click to view available options
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Para análisis de ultratrazas, reactivo de reducción
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Iron
Punto de fusión 1535.0°C
Punto de ebullición 3000.0°C
Intervalo de porcentaje de ensayo 99.99%
Cantidad 5 g
Formula Weight (peso de la fórmula) 55.84
Porcentaje de pureza 99.99%
Grado base de trazas de metales
Envase Frasco de vidrio
Formulario Gránulos