missing translation for 'onlineSavingsMsg'
Learn More

IRF5 Antibody (10T1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-04307AF405

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213195

  • 558.30€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

IRF5 Monoclonal specifically detects IRF5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

IRF5
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
IRF5
Recombinant human IRF5 (176-240 aa) purified from E. coli
0.1 mL
Apoptosis, Death Receptor Signaling Pathway
3663
Human, Mouse
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
10T1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Mouse
Protein G purified
RUO
Primary
Anti-human IRF-5 mAb, clone 10T1, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF-5 (176-240 aa).
Store at 4C in the dark.
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales