missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQCF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549327
Detalles adicionales : Peso : 0.01000kg
Descripción
IQCF6 Polyclonal antibody specifically detects IQCF6 in Human samples. It is validated for Western Blot.Especificaciones
IQCF6 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
A8MYZ5 | |
IQCF6 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human IQCF6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
IQCF6 | |
IQ domain-containing protein F6; IQ motif containing F6; IQCF6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
440956 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |