Learn More
Mettler Toledo™ Ion Selective Electrode DX264-Cu Copper
Product Code. 11967701
Change view
Click to view available options
pH Range:
2 to 8
Unit Size:
Each
Product Code. pH Range Unit Size
11967701 2 to 8 Each
1 options
This item is not returnable. View return policy
Product Code. 11967701 Supplier Mettler Toledo™ Supplier No. 51107678
  • ONLINE EXCLUSIVE PRICE
    1505.00€
    1299.21€ / paquete de 1
    Ahorro 205.79€
    13% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type DX264-Cu Copper half-cell
Material POM
pH Range 2 to 8
Temperature (Metric) Range 0°C to 80°C