missing translation for 'onlineSavingsMsg'
Learn More

Bromide Standard for Ion Chromatography, 1000μg/mL Br-, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Marca:  SPEX Certiprep AS-BR9-2X

Detalles adicionales : Peso : 0.59000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Cantidad. 500mL
Envase Triple-leached LDPE bottle

 Ver más versiones de este producto

Código de producto. 11495908

  • 108.00€ / 500 ml


Fecha de envío estimada
del proveedor 19-02-2019
Añadir a la cesta

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Descripción

Descripción

AnalyteConcentration
Br-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Especificación

Identificadores de productos químicos

7732-18-5

Especificación

Bromide Standard
99.9
1000μg/mL Br−
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
7732-18-5
99.9
Triple-leached LDPE bottle
H2O
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Videos

Product intended for laboratory use only