missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride Solution, Honeywell™ Fluka™

Catalog Number p-7120486
Click to view available options
Cantidad:
100 mL
800 mL
Envase:
Glass Bottle
Plastic Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
7790-99-0
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
24640
iodochlorane
ClI
Este artículo no se puede devolver. Vea la política de devoluciones
7790-99-0
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
24640
iodochlorane
ClI

1.0 M in methylene chloride

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Iodine Monochloride Solution
Forma física Liquid
Cantidad 100 mL
Fórmula lineal ICl
Número UN UN3264
Beilstein 3587194
Formula Weight (peso de la fórmula) 162.36g/mol
Envase Plastic Bottle
Densidad 1.42g/mL at 25°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'