missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Monochloride Solution, Honeywell™ Fluka™
Click to view available options
Cantidad:
100 mL
800 mL
Envase:
Glass Bottle
Plastic Bottle
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Iodine Monochloride Solution |
Forma física | Liquid |
Cantidad | 800 mL |
Fórmula lineal | ICl |
Número UN | UN3264 |
Beilstein | 3587194 |
Formula Weight (peso de la fórmula) | 162.36g/mol |
Envase | Glass Bottle |
Densidad | 1.42g/mL at 25°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'