Learn More
Yoduro, solución estándar para cromatografía iónica, Specpure™, I- 1000 μg/ml, Thermo Scientific Chemicals
Catalog Number 11316119
Click to view available options
Cantidad:
100 mL
500 mL
MFCD00011355
MFCD00011355
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Iodide, Ion chromatography standard solution
Nombre de nota I- 1000μg/mL
Cantidad 500 mL
Grado Specpure™
Forma física Líquida