missing translation for 'onlineSavingsMsg'
Learn More

INVS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10585-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817063

  • 129.00€ / 20 microlitros
Fecha estimada de envío: 13-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is a rabbit polyclonal antibody raised against part of N-terminus of human INVS. The calculated molecular weight of INVS is 80 kDa, but the modified INVS protein is about 100-110 kDa.

This gene encodes a protein containing multiple ankyrin domains and two IQ calmodulin-binding domains. The encoded protein may function in renal tubular development and function, and in left-right axis determination. This protein interacts with nephrocystin and infers a connection between primary cilia function and left-right axis determination. A similar protein in mice interacts with calmodulin. Mutations in this gene have been associated with nephronophthisis type 2. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Especificaciones

Especificaciones

INVS
Polyclonal
Unconjugated
INVS
INV, inversin, INVS, KIAA0573, Nephrocystin 2
Rabbit
Antigen Affinity Chromatography
RUO
16348, 27130, 313228
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O89019, Q9Y283
Invs
INVS Fusion Protein Ag0934
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales