Learn More
Invitrogen™ Yeast Homozygous Diploid Plate
Product Code. 10747245
Change view
Click to view available options
Format:
Individual Plates
Unit Size:
Each
Product Code. Format Unit Size
10747245 Individual Plates Each
1 options
This item is not returnable. View return policy
Product Code. 10747245 Supplier Invitrogen™ Supplier No. 95401.H1P
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Product Type Yeast Homozygous Diploid Plate
Libraries Yeast deletion library created by collaboration of 8 North American and 8 European labs composing the Saccharomyces Genome Deletion Project
Culture Type Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)
Format Individual Plates