missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SuperFi™ Green DNA Polymerase

Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps

Marca:  Invitrogen™ 12357010

Detalles adicionales : Peso : 0.04500kg

 Ver más versiones de este producto

Código de producto. 15585033

  • 201.00€ / 100 unidades


En stock, listo para ser enviado: 3
Añadir a la cesta

Descripción

Descripción

  • Error-free PCR products due to exceptional >100X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content 
  • Convenient workflow with room temperature reaction setup and direct gel loading with green buffer format
Especificaciones

Especificaciones

• 50 µL Platinum SuperFi DNA Polymerase (2U/µL)
• 2 x 1.25 mL SuperFi Green Buffer (5X)
• 1.25 mL SuperFi GC Enhancer (5X)

Store at -5 to -30°C.
Built-In Hot Start
Approved for shipment on Wet or Dry Ice
Fast
100 Units
100 X
Platinum™, SuperFi™
Platinum SuperFi DNA Polymerase
Blunt
Videos
Documentos