missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16455-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2.Especificaciones
INTS12 | |
Polyclonal | |
Unconjugated | |
INTS12 | |
INT12, integrator complex subunit 12, INTS12, PHD finger protein 22, PHF22, SBBI22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295448, 57117, 71793 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FR3, Q96CB8, Q9D168 | |
Ints12 | |
INTS12 Fusion Protein Ag9403 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |