missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7 Antibody (2G18), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NB110-94147PECY7

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213755

  • 587.46€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

Integrin beta 7
Monoclonal
PE-Cyanine7
PBS with 0.05% Sodium Azide
ITGB7
Cell lysates of mouse lymphocytes
0.1 mL
Primary
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC.
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
2G18
Flow Cytometry
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Rat
Protein A or G purified
Signal Transduction
3695.0
Mouse
IgG2
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales