Learn More
Integrin beta 4/CD104 Antibody (439-9B), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Marca:  Bio-Techne NBP1-43369FR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215894
  • 519.42€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
Integrin beta 4/CD104
Monoclonal
DyLight 680
P16144
ITGB4
The immunogen for this antibody was CD104.
0.1 mL
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
3691
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
439-9B
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
Rat
Affinity Purified
RUO
Primary
Human, Bovine
IgG2b