Learn More
Novus Biologicals™ Integrin beta 2/CD18 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-12072
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216043
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
The Integrin beta 2/CD18 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC21126 and based on accession number NM_211. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens integrin, beta 2 (complement component 3 receptor 3 and 4 subunit) (ITGB2), transcript variant 1, mRNA.
Native