Learn More
Integrin beta 1/CD29 Antibody (MEM-101A), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB500-383UV
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215130
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Porcine, Canine, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones
Especificaciones
Integrin beta 1/CD29
Monoclonal
DyLight 350
P05556
ITGB1
Raji Burkitt's lymphoma cell line Raji (NM_002211.3).
0.1 mL
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
3688
Human, Porcine, Canine, Mouse (Negative)
Purified
Western Blot, Flow Cytometry
MEM-101A
Western Blot, Flow Cytometry
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Mouse
Protein A purified
RUO
Primary
The antibody MEM-101A reacts with CD29 (integrin beta1 chain), a 130 kDa single chain type I glycoprotein expressed as a heterodimer (non-covalently associated with the integrin alpha subunits 1-6). CD29 is broadly expressed on majority of hematopoietic and non-hematopoietic cells (leukocytes, platelets, fibroblasts, endothelial cells, epithelial cells and mast cells). HLDA VI; WS Code AS A048
Store at 4C in the dark.
IgG1