Learn More
Integrin alpha 2b/CD41 Antibody (7E3 (Abciximab)), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Bio-Techne NBP2-81051IR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214169
  • 588.54€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
Integrin alpha 2b/CD41
Monoclonal
DyLight 755
50mM Sodium Borate
ITGA2B
CD41 is the integrin alpha chain 2b forming a fibrinogen receptor in heterodimeric complex with beta 3.
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
3674
Human
Purified
Flow Cytometry
7E3 (Abciximab)
Flow Cytometry
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Rabbit
Protein A purified
RUO
Primary
Recognises human Integrin alpha 2b/CD41.
Store at 4°C in the dark.
IgG κ