missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Pyrophosphatase/PPA1 Antibody (4G4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-59471H

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214320

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Western Blot, ELISA
Especificaciones

Especificaciones

Inorganic Pyrophosphatase/PPA1
Monoclonal
HRP
diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061
Recombinant human Inorganic Pyrophosphatase/PPA1 (1-289aa) purified from E. coli
0.1 mL
metabolism, Signal Transduction
5464
Store at 4°C in the dark.
IgG2b λ
Western Blot, ELISA
4G4
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales